Assay Details
| Gene Symbol: tpi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114234.1 | NP_001107706.1 | 1 - 2 | 145 | 76 | |
| GenBank mRNA | BC158276.1 | - | 1 - 2 | 145 | 76 | |
| CR942691.2 | - | 1 - 2 | 138 | 76 | ||
More Information
Set Membership: |
|

tpi1
Genomic Map