Assay Details
Gene Symbol: apoo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114243.1 | NP_001107715.1 | 6 - 7 | 484 | 79 | |
XM_012956316.1 | XP_012811770.1 | 6 - 7 | 501 | 79 | ||
XM_012956317.2 | XP_012811771.1 | 6 - 7 | 539 | 79 | ||
GenBank mRNA | BC135301.1 | - | 5 - 6 | 469 | 79 | |
BC155970.1 | - | 6 - 7 | 484 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
