Assay Details
| Gene Symbol: arl6ip4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114257.1 | NP_001107729.1 | 2 - 3 | 285 | 72 | |
| XM_012954493.2 | XP_012809947.1 | 2 - 3 | 774 | 72 | ||
| GenBank mRNA | BC158419.1 | - | 2 - 3 | 285 | 72 | |
| BC161359.1 | - | 2 - 3 | 285 | 72 | ||
More Information
Set Membership: |
|

arl6ip4
Genomic Map