Assay Details
Gene Symbol: rtn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114267.2 | NP_001107739.1 | 4 - 5 | 737 | 65 | |
GenBank mRNA | CU025177.1 | - | 4 - 5 | 641 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
