Assay Details
| Gene Symbol: commd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122721.1 | NP_001116193.1 | 1 - 2 | 209 | 89 | |
| GenBank mRNA | BC118890.1 | - | 1 - 2 | 152 | 89 | |
| BC160794.1 | - | 1 - 2 | 209 | 89 | ||
| CT027909.1 | - | 1 - 2 | 189 | 89 | ||
More Information
Set Membership: |
|

commd1
Genomic Map