Assay Details
Gene Symbol: pum3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122795.1 | NP_001116267.1 | 2 - 3 | 311 | 63 | |
GenBank mRNA | BC135310.1 | - | 2 - 3 | 260 | 63 | |
BC161436.1 | - | 2 - 3 | 311 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
