Assay Details
| Gene Symbol: bves | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123416.1 | NP_001116888.1 | 2 - 3 | 363 | 64 | |
| XM_012962694.2 | XP_012818148.1 | 2 - 3 | 503 | 64 | ||
| GenBank mRNA | BC160594.1 | - | 2 - 3 | 363 | 64 | |
More Information
Set Membership: |
|

bves
Genomic Map