Assay Details
Gene Symbol: efna2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123420.1 | NP_001116892.1 | 3 - 4 | 983 | 70 | |
GenBank mRNA | BC161541.1 | - | 3 - 4 | 983 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
