Assay Details
| Gene Symbol: adrb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123425.2 | NP_001116897.2 | 1 | 1015 | 93 | |
| GenBank mRNA | BC160515.1 | - | 1 | 949 | 93 | |
| DT425584.1 | - | 1 | 647 | 93 | ||
| EL798180.1 | - | 1 | 651 | 93 | ||
More Information
Set Membership: |
|

adrb2
Genomic Map