Assay Details
| Gene Symbol: rnf19a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123427.1 | NP_001116899.1 | 8 - 9 | 1839 | 62 | |
| XM_018094531.1 | XP_017950020.1 | 9 - 10 | 2023 | 62 | ||
| XM_018094532.1 | XP_017950021.1 | 8 - 9 | 1864 | 62 | ||
| GenBank mRNA | BC160518.1 | - | 8 - 9 | 1839 | 62 | |
More Information
Set Membership: |
|

rnf19a
Genomic Map