Assay Details
| Gene Symbol: unkl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123443.1 | NP_001116915.1 | 1 - 2 | 216 | 60 | |
| XM_012970032.2 | XP_012825486.1 | 1 - 2 | 1637 | 60 | ||
| GenBank mRNA | BC160454.1 | - | 1 - 2 | 216 | 60 | |
More Information
Set Membership: |
|

unkl
Genomic Map