Assay Details
Gene Symbol: cd151 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123447.1 | NP_001116919.1 | 6 - 7 | 717 | 80 | |
XM_012960584.2 | XP_012816038.1 | 7 - 8 | 829 | 80 | ||
XM_012960585.2 | XP_012816039.1 | 7 - 8 | 913 | 80 | ||
XM_018092916.1 | XP_017948405.1 | 8 - 9 | 936 | 80 | ||
GenBank mRNA | BC161493.1 | - | 6 - 7 | 717 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
