Assay Details
Gene Symbol: cdkn2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123460.1 | NP_001116932.1 | 2 - 3 | 558 | 73 | |
GenBank mRNA | BC160579.1 | - | 2 - 3 | 558 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
