Assay Details
Gene Symbol: fhl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123485.1 | NP_001116957.1 | 4 - 5 | 714 | 73 | |
GenBank mRNA | BC160566.1 | - | 4 - 5 | 714 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
