Assay Details
| Gene Symbol: rrm2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126501.1 | NP_001119973.1 | 2 - 3 | 190 | 62 | |
| GenBank mRNA | BC158190.1 | - | 2 - 3 | 190 | 62 | |
More Information
Set Membership: |
|

rrm2b
Genomic Map