Assay Details
Gene Symbol: rpl10a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126557.1 | NP_001120029.1 | 1 - 2 | 544 | 66 | |
XM_012955646.1 | XP_012811100.1 | 2 - 3 | 399 | 66 | ||
XM_012955647.1 | XP_012811101.1 | 1 - 2 | 87 | 66 | ||
GenBank mRNA | BC135694.1 | - | 1 - 2 | 77 | 66 | |
BC158335.1 | - | 1 - 2 | 544 | 66 | ||
BC168113.1 | - | 1 - 2 | 72 | 66 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_012955647.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
