Assay Details
Gene Symbol: shpk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126606.1 | NP_001120078.1 | 3 - 4 | 493 | 74 | |
XM_012956440.1 | XP_012811894.1 | 3 - 4 | 418 | 74 | ||
XM_012956441.2 | XP_012811895.1 | 3 - 4 | 413 | 74 | ||
XM_018091083.1 | XP_017946572.1 | 3 - 4 | 414 | 74 | ||
GenBank mRNA | BC158517.1 | - | 3 - 4 | 493 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
