Assay Details
Gene Symbol: grina | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126613.1 | NP_001120085.1 | 6 - 7 | 625 | 59 | |
XM_018094536.1 | XP_017950025.1 | 6 - 7 | 1250 | 59 | ||
GenBank mRNA | BC158526.1 | - | 6 - 7 | 625 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
