Assay Details
| Gene Symbol: pex19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126617.1 | NP_001120089.1 | 5 - 6 | 591 | 63 | |
| GenBank mRNA | BC158532.1 | - | 5 - 6 | 591 | 63 | |
More Information
Set Membership: |
|

pex19
Genomic Map