Assay Details
| Gene Symbol: mrpl15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126635.1 | NP_001120107.1 | 1 - 2 | 216 | 61 | |
| GenBank mRNA | BC158958.1 | - | 1 - 2 | 216 | 61 | |
More Information
Set Membership: |
|

mrpl15
Genomic Map