Assay Details
| Gene Symbol: vps35l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012970827.2 | XP_012826281.1 | 6 - 7 | 578 | 69 | |
| XM_012970828.2 | XP_012826282.1 | 6 - 7 | 578 | 69 | ||
| GenBank mRNA | BC159055.1 | - | 6 - 7 | 525 | 69 | |
| CT025264.2 | - | 6 - 7 | 506 | 69 | ||
More Information
Set Membership: |
|

vps35l
Genomic Map