Assay Details
Gene Symbol: arf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126681.1 | NP_001120153.1 | 3 - 4 | 364 | 67 | |
XM_012956457.2 | XP_012811911.1 | 3 - 4 | 397 | 67 | ||
GenBank mRNA | BC159149.1 | - | 3 - 4 | 364 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
