Assay Details
Gene Symbol: tesk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126723.2 | NP_001120195.2 | 439 | 66 | ||
GenBank mRNA | BC159315.1 | - | 263 | 66 | ||
CX363373.2 | - | 433 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
