Assay Details
| Gene Symbol: etv2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126744.2 | NP_001120216.2 | 4 - 5 | 702 | 70 | |
| XM_012966060.2 | XP_012821514.1 | 4 - 5 | 851 | 70 | ||
| GenBank mRNA | BC159381.1 | - | 4 - 5 | 689 | 70 | |
More Information
Set Membership: |
|

etv2
Genomic Map