Assay Details
Gene Symbol: ikbkg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126749.1 | NP_001120221.1 | 2 - 3 | 262 | 84 | |
XM_012960604.2 | XP_012816058.1 | 2 - 3 | 311 | 84 | ||
XM_012960605.2 | XP_012816059.1 | 2 - 3 | 435 | 84 | ||
XM_012960606.2 | XP_012816060.1 | 2 - 3 | 307 | 84 | ||
XM_012960607.2 | XP_012816061.1 | 2 - 3 | 435 | 84 | ||
XM_018096158.1 | XP_017951647.1 | 2 - 3 | 294 | 84 | ||
GenBank mRNA | BC159400.1 | - | 2 - 3 | 262 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
