Assay Details
| Gene Symbol: map6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126750.1 | NP_001120222.1 | 2 - 3 | 1075 | 97 | |
| XM_012956469.2 | XP_012811923.1 | 2 - 3 | 1010 | 97 | ||
| XM_012956471.2 | XP_012811925.1 | 2 - 3 | 1010 | 97 | ||
| XM_012956472.2 | XP_012811926.1 | 2 - 3 | 3044 | 97 | ||
| GenBank mRNA | BC159130.1 | - | 2 - 3 | 1075 | 97 | |
More Information
Set Membership: |
|

map6
Genomic Map