Assay Details
| Gene Symbol: nab1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126751.1 | NP_001120223.1 | 6 - 7 | 1457 | 77 | |
| XM_012968009.2 | XP_012823463.1 | 5 - 6 | 1507 | 77 | ||
| GenBank mRNA | BC160386.1 | - | 6 - 7 | 1457 | 77 | |
More Information
Set Membership: |
|

nab1
Genomic Map