Assay Details
| Gene Symbol: pea15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126828.1 | NP_001120300.1 | 3 - 4 | 543 | 80 | |
| GenBank mRNA | BC160637.1 | - | 3 - 4 | 543 | 80 | |
More Information
Set Membership: |
|

pea15
Genomic Map