Assay Details
| Gene Symbol: dyrk1a.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126901.1 | NP_001120373.1 | 5 - 6 | 958 | 93 | |
| XM_012966071.2 | XP_012821525.1 | 4 - 5 | 93 | |||
| XM_012966072.2 | XP_012821526.1 | 4 - 5 | 93 | |||
| XM_012966073.2 | XP_012821527.1 | 5 - 6 | 93 | |||
| XM_012966074.2 | XP_012821528.1 | 4 - 5 | 93 | |||
| XM_012966075.2 | XP_012821529.1 | 5 - 6 | 93 | |||
| XM_012966076.2 | XP_012821530.1 | 4 - 5 | 93 | |||
| GenBank mRNA | BC161066.1 | - | 5 - 6 | 958 | 93 | |
| CT030289.1 | - | 5 - 6 | 93 | |||
More Information
Set Membership: |
|

dyrk1a.2
Genomic Map