Assay Details
Gene Symbol: adra2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126903.1 | NP_001120375.1 | 1 | 1555 | 94 | |
GenBank mRNA | BC161069.1 | - | 1 | 1555 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
