Assay Details
| Gene Symbol: use1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127019.1 | NP_001120491.1 | 7 - 8 | 606 | 67 | |
| XM_012956366.2 | XP_012811820.1 | 6 - 7 | 716 | 67 | ||
| GenBank mRNA | BC161367.1 | - | 7 - 8 | 606 | 67 | |
More Information
Set Membership: |
|

use1
Genomic Map