Assay Details
| Gene Symbol: hsd3b7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127029.1 | NP_001120501.1 | 4 - 5 | 449 | 99 | |
| XM_012954311.2 | XP_012809765.1 | 3 - 4 | 441 | 99 | ||
| XM_018097043.1 | XP_017952532.1 | 4 - 5 | 497 | 99 | ||
| GenBank mRNA | BC161389.1 | - | 4 - 5 | 449 | 99 | |
More Information
Set Membership: |
|

hsd3b7
Genomic Map