Assay Details
| Gene Symbol: slc10a7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004977.2 | NP_001004977.2 | 5 - 6 | 545 | 63 | |
| GenBank mRNA | CR760903.2 | - | 5 - 6 | 545 | 63 | |
More Information
Set Membership: |
|

slc10a7
Genomic Map