Assay Details
| Gene Symbol: gpx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015740.3 | NP_001015740.2 | 1 - 2 | 246 | 82 | |
| GenBank mRNA | BC089675.1 | - | 1 - 2 | 213 | 82 | |
| BC121514.1 | - | 1 - 2 | 167 | 82 | ||
| BC157770.1 | - | 1 - 2 | 212 | 82 | ||
| CR760260.2 | - | 1 - 2 | 205 | 82 | ||
| EG655675.1 | - | 1 - 2 | 285 | 82 | ||
More Information
Set Membership: |
|

gpx1
Genomic Map