Assay Details
Gene Symbol: capza1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017324.3 | NP_001017324.1 | 1 - 2 | 117 | 63 | |
GenBank mRNA | BC067322.1 | - | 46 | 63 | ||
BC158140.1 | - | 1 - 2 | 117 | 63 | ||
CR848613.2 | - | 1 - 2 | 65 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
