Assay Details
Gene Symbol: cmtm6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039741.1 | NP_001034830.1 | 2 - 3 | 436 | 70 | |
GenBank mRNA | BC090802.1 | - | 2 - 3 | 428 | 70 | |
BC167269.1 | - | 2 - 3 | 680 | 70 | ||
CR855776.2 | - | 2 - 3 | 436 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
