Assay Details
Gene Symbol: fkbp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114204.1 | NP_001107676.1 | 1 - 2 | 308 | 83 | |
GenBank mRNA | BC087818.1 | - | 1 - 2 | 244 | 83 | |
BC157309.1 | - | 1 - 2 | 308 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
