Assay Details
Gene Symbol: ovca2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114239.1 | NP_001107711.1 | 1 - 2 | 191 | 104 | |
GenBank mRNA | BC135890.1 | - | 1 - 2 | 185 | 104 | |
BC153671.1 | - | 1 - 2 | 191 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
