Assay Details
Gene Symbol: hyal2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126514.1 | NP_001119986.1 | 3 - 4 | 1289 | 94 | |
GenBank mRNA | BC158239.1 | - | 3 - 4 | 1289 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
