Assay Details
Gene Symbol: tspan9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126530.1 | NP_001120002.1 | 9 - 10 | 997 | 68 | |
XM_012958533.2 | XP_012813987.1 | 6 - 7 | 1293 | 68 | ||
XM_012958535.2 | XP_012813989.1 | 7 - 8 | 1176 | 68 | ||
XM_012958536.2 | XP_012813990.1 | 9 - 10 | 1135 | 68 | ||
XM_012958537.2 | XP_012813991.1 | 8 - 9 | 1086 | 68 | ||
XM_018091932.1 | XP_017947421.1 | 8 - 9 | 1082 | 68 | ||
GenBank mRNA | BC158275.1 | - | 9 - 10 | 997 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
