Assay Details
Gene Symbol: fam228b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126582.1 | NP_001120054.1 | 5 - 6 | 401 | 93 | |
XM_012970053.1 | XP_012825507.1 | 5 - 6 | 654 | 93 | ||
XM_012970054.1 | XP_012825508.1 | 3 - 4 | 938 | 93 | ||
XM_012970055.2 | XP_012825509.1 | 3 - 4 | 289 | 93 | ||
XM_018093782.1 | XP_017949271.1 | 5 - 6 | 654 | 93 | ||
GenBank mRNA | BC158429.1 | - | 5 - 6 | 401 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
