Assay Details
Gene Symbol: fam228b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126582.1 | NP_001120054.1 | 6 - 7 | 611 | 80 | |
XM_012970053.1 | XP_012825507.1 | 6 - 7 | 864 | 80 | ||
XM_012970054.1 | XP_012825508.1 | 4 - 5 | 1148 | 80 | ||
XM_012970055.2 | XP_012825509.1 | 4 - 5 | 499 | 80 | ||
GenBank mRNA | BC158429.1 | - | 6 - 7 | 611 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
