Assay Details
Gene Symbol: c1orf43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126591.1 | NP_001120063.1 | 5 - 6 | 512 | 74 | |
GenBank mRNA | BC158474.1 | - | 5 - 6 | 512 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
