Assay Details
Gene Symbol: lrp2bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126597.1 | NP_001120069.1 | 5 - 6 | 580 | 72 | |
XM_012955694.2 | XP_012811148.1 | 5 - 6 | 528 | 72 | ||
XM_018090641.1 | XP_017946130.1 | 5 - 6 | 862 | 72 | ||
GenBank mRNA | BC158486.1 | - | 5 - 6 | 580 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
