Assay Details
Gene Symbol: arl3l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126745.2 | NP_001120217.2 | 2 - 3 | 186 | 63 | |
NM_001345943.1 | NP_001332872.1 | 2 - 3 | 434 | 63 | ||
XM_018091935.1 | XP_017947424.1 | 3 - 4 | 1957 | 63 | ||
GenBank mRNA | AL677452.2 | - | 2 - 3 | 434 | 63 | |
BX686615.1 | - | 4 - 5 | 542 | 63 | ||
CX365546.2 | - | 2 - 3 | 187 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
