Assay Details
Gene Symbol: arl3l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126745.2 | NP_001120217.2 | 4 - 5 | 349 | 79 | |
NM_001345943.1 | NP_001332872.1 | 4 - 5 | 597 | 79 | ||
XM_018091935.1 | XP_017947424.1 | 5 - 6 | 2120 | 79 | ||
GenBank mRNA | BX686615.1 | - | 2 - 3 | 379 | 79 | |
CX333591.2 | - | 3 - 4 | 239 | 79 | ||
CX365546.2 | - | 4 - 5 | 350 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() |
