Assay Details
Gene Symbol: stambp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126765.2 | NP_001120237.1 | 4 - 5 | 753 | 61 | |
XM_012958567.2 | XP_012814021.1 | 5 - 6 | 892 | 61 | ||
XM_012958568.2 | XP_012814022.1 | 5 - 6 | 835 | 61 | ||
GenBank mRNA | BC160416.1 | - | 4 - 5 | 755 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
