Assay Details
| Gene Symbol: ttc33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126771.1 | NP_001120243.1 | 2 - 3 | 292 | 90 | |
| XM_012955907.2 | XP_012811361.1 | 2 - 3 | 452 | 90 | ||
| XM_012955908.2 | XP_012811362.1 | 2 - 3 | 349 | 90 | ||
| XM_018090842.1 | XP_017946331.1 | 2 - 3 | 344 | 90 | ||
| XM_018090847.1 | XP_017946336.1 | 2 - 3 | 343 | 90 | ||
| GenBank mRNA | BC160438.1 | - | 2 - 3 | 292 | 90 | |
More Information
Set Membership: |
|

ttc33
Genomic Map