Assay Details
Gene Symbol: rpain | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126782.1 | NP_001120254.1 | 1 - 2 | 90 | 98 | |
GenBank mRNA | BC160480.1 | - | 1 - 2 | 90 | 98 | |
CR761991.2 | - | 1 - 2 | 82 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
