Assay Details
Gene Symbol: psmg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126789.1 | NP_001120261.1 | 2 - 3 | 262 | 71 | |
GenBank mRNA | BC160506.1 | - | 2 - 3 | 262 | 71 | |
CR942702.2 | - | 2 - 3 | 250 | 71 | ||
CU024917.1 | - | 2 - 3 | 258 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
